missing translation for 'onlineSavingsMsg'
Learn More

(R)-Filanesib, MedChemExpress

Product Code. 16452467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452467 50 mg 50mg
16416666 100 mg 100mg
16416358 10 mg 10mg
16408947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452467 Supplier Medchem Express Supplier No. HY15187A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM.

(R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM.

Specifications

Chemical Name or Material (R)-Filanesib
CAS 885060-08-2
Color White
Purity Grade Notes Research
Molecular Formula C20H22F2N4O2S
Quantity 50 mg
Synonym (R)-ARRY-520
Solubility Information DMSO : ≥ 100 mg/mL (237.82 mM)
SMILES O=C(N1[C@](C2=CC=CC=C2)(CCCN)SC(C3=CC(F)=CC=C3F)=N1)N(OC)C
Molecular Weight (g/mol) 420.48
Formula Weight 420.48
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.