missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Fesoterodine Fumarate, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Fesoterodine Fumarate |
| CAS | 286930-03-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C26 H37 N O3 . C4 H4 O4 |
| Quantity | 25 mg |
| Synonym | 2-Methyl-propanoic acid 2-[(1R)-3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-(hydroxymethyl)phenyl ester (2E)-2-butenedioate (1:1), Propanoic acid, 2-methyl-, 2-[(1R)-3-[bis(1-methylethyl)amino]-1-phenylpropyl]-4-(hydroxymethyl)phenyl ester, (2E)-2-butenedioate (1:1) (salt) (9CI), Fesoterodine fumarate, SMP 8272, SPM 907, Toviaz |
| InChI Formula | InChI=1S/C26H37NO3.C4H4O4/c1-18(2)26(29)30-25-13-12-21(17-28)16-24(25)23(22-10-8-7-9-11-22)14-15-27(19(3)4)20(5)6;5-3(6)1-2-4(7)8/h7-13,16,18-20,23,28H,14-15,17H2,1-6H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t23-;/m1./s1 |
| SMILES | CC(C)N(CC[C@H](c1ccccc1)c2cc(CO)ccc2OC(=O)C(C)C)C(C)C.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;[2-[(1R)-3-[di(propan-2-yl)amino]-1-phenylpropyl]-4-(hydroxymethyl)phenyl] 2-methylpropanoate |
| Molecular Weight (g/mol) | 527.65 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?