missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Epichlorohydrin, 98+%

Product Code. 11379045
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379045 25 g 25g
11359045 1 g 1g
11369045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379045 Supplier Thermo Scientific Alfa Aesar Supplier No. L14198.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51594-55-9 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51594-55-9
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077759
InChI Key BRLQWZUYTZBJKN-VKHMYHEASA-N
Synonym r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
PubChem CID 2734062
ChEBI CHEBI:18662
IUPAC Name (2R)-2-(chloromethyl)oxirane
SMILES ClC[C@H]1CO1

Specifications

Density 1.18
Boiling Point 116°C to 117°C
Flash Point 32°C (90°F)
Refractive Index 1.438
Quantity 25 g
UN Number UN2023
Beilstein 1420785
Optical Rotation −35° (c=1 in Methanol)
Formula Weight 92.53
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Epichlorohydrin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.