missing translation for 'onlineSavingsMsg'
Learn More

(R)-Diphenyl (((1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonate, TRC

Product Code. 30378864
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378864 250 mg 250mg
30388449 100 mg 100mg
30386740 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378864 Supplier Toronto Research Chemicals Supplier No. TRCD431035250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 342631-41-8
Molecular Formula C21H22N5O4P
Molecular Weight (g/mol) 439.4
Synonym [(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid, Diphenyl Ester
SMILES C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(Oc3ccccc3)Oc4ccccc4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H22N5O4P/c1-16(12-26-14-25-19-20(22)23-13-24-21(19)26)28-15-31(27,29-17-8-4-2-5-9-17)30-18-10-6-3-7-11-18/h2-11,13-14,16H,12,15H2,1H3,(H2,22,23,24)/t16-/m1/s1
Formula Weight 439.1409
Chemical Name or Material (R)-Diphenyl (((1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.