missing translation for 'onlineSavingsMsg'
Learn More

(R)-Desmethyl Citalopram Hydrochloride, TRC

Product Code. 30010336
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010336 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010336 Supplier Toronto Research Chemicals Supplier No. TRCD2915651MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144010-85-5
Molecular Formula C19 H19 F N2 O . Cl H
Quantity 1 mg
Synonym (R)-Desmethylcitalopram Hydrochloride, (R)-Desmethyl Citalopram Hydrochloride, (1R)-1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzofurancarbonitrile hydrochloride
InChI Formula InChI=1S/C19H19FN2O.ClH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H/t19-;/m1./s1
SMILES Cl.CNCCC[C@@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC Name (1R)-1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrochloride
Molecular Weight (g/mol) 346.83
Formula Weight 346.1248
Chemical Name or Material (R)-Desmethyl Citalopram Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.