missing translation for 'onlineSavingsMsg'
Learn More

R-(-)-Deprenyl Hydrochloride, TRC

Product Code. 30132444
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132444 5 g 5g
18040930 100 mg 100mg
18062230 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132444 Supplier Toronto Research Chemicals Supplier No. TRCD2886415G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14611-52-0
Molecular Formula C13 H17 N . Cl H
Molecular Weight (g/mol) 223.74
Synonym Selegiline Hydrochloride, Benzeneethanamine, N,α-dimethyl-N-2-propyn-1-yl-, hydrochloride (1:1), (αR)-, Benzeneethanamine, N,α-dimethyl-N-2-propynyl-, hydrochloride, (R)-, Benzeneethanamine, N,α-dimethyl-N-2-propynyl-, hydrochloride, (αR)- (9CI), (-)-Deprenil hydrochloride, (-)-Deprenyl hydrochloride, Eldeprine, Eldepryl, FP Tablet 2.5, Jumex, Movergan, Plurimen, R-Selegiline hydrochloride, Tonus, l-Deprenyl hydrochloride
IUPAC Name (2R)-N-methyl-1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride
SMILES Cl.C[C@H](Cc1ccccc1)N(C)CC#C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H17N.ClH/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12H,10-11H2,2-3H3;1H/t12-;/m1./s1
Formula Weight 223.1128
Percent Purity >95%
Chemical Name or Material Selegiline Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.