missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Deprenyl hydrochloride

Product Code. 15483899
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483899 250 mg 250mg
15473899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483899 Supplier Thermo Scientific Alfa Aesar Supplier No. J61286.MD
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective MAO-B inhibitor; anti-Parkinsonian agent | CAS: 14611-52-0 | C13H18ClN | 223.74 g/mol

(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

Solubility
Soluble in water at 100mM

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14611-52-0
Molecular Formula C13H18ClN
Molecular Weight (g/mol) 223.74
MDL Number MFCD00069299
InChI Key IYETZZCWLLUHIJ-UTONKHPSSA-N
Synonym selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
PubChem CID 26758
ChEBI CHEBI:9087
IUPAC Name hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
SMILES [H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C

Specifications

Color White
Quantity 250 mg
Merck Index 14,8428
Solubility Information Soluble in water at 100mM
Optical Rotation −12° (c=1 in Water)
Formula Weight 223.74
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Deprenyl hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.