missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Zebrafish BMP-2a Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£380.12
Specifications
Gene ID (Entrez) | 30631 |
---|---|
Molecular Weight (g/mol) | 13 kDa (monomer) |
Name | BMP-2a |
Quantity | 10μg |
Source | E. coli-derived zebrafish BMP-2a protein Gln272-Arg386,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18319667
|
R&D Systems™
111-BM-010/CF |
£380.12
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18187160
|
R&D Systems™
111-BM-010 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£380.12
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Zebrafish BMP-2a Protein is derived from E. coli. The Recombinant Zebrafish BMP-2a Protein has been validated for the following applications: Bioactivity.Specifications
30631 | |
BMP-2a | |
E. coli-derived zebrafish BMP-2a protein Gln272-Arg386,with an N-terminal Met | |
Disulfide-linked homodimer | |
Recombinant |
13 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |