missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant West Nile Virus NS3 Protease Protein, CF

Product Code. 18320268
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320268 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320268 Supplier R&D Systems™ Supplier No. 2907SE020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate pERTKR-AMC (Catalog # ES013 ). The specific activity is >750 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number YP_001527877.1
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 912267
Molecular Weight (g/mol) Observed M.W.: 32 kDa and 18-22 kDa, under reducing conditions.
Name wnvNS3 Protease
Quantity 20 μg
Source E. coli-derived viral wnvNS3 Protease proteinMHHHHHH NS2b (Ser1423-Lys1470) (Ser1435Thr) (N-terminus) Accession # YP_001527877.1 GGGGSGGGG West Nile Virus NS3 (Gly1506-Leu1689 Accession # YP_001527877.1 GGGGSGGGG(C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol flavivirus polyprotein gene
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile, deionized water.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein wnvNS3 Protease
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.