missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-G 1008
GREENER_CHOICE

Product Code. 18730528
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18730528

Brand: Tocris Bioscience™ 5355/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective GPR39 agonist

Potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.

TC-G 1008 is a potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TC-G 1008
CAS 1621175-65-2
Synonym N-[3-Chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]methanesulfonate
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C18H19ClN6O2S
Formula Weight 418.9
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.