Learn More
R&D Systems™ TACS Nuclease and Buffer
Product Code. 18789266
Click to view available options
Quantity:
15 μL
Unit Size:
15µL
Product Code. 18789266
Brand: R&D Systems™ 480030N
Highly purified recombinant endonuclease specific for all forms of DNA
  • TACS-Nuclease has high specific activity, is tolerant to a variety of buffer conditions, and is stable at both 4°C and at -20°C
  • TACS-Nuclease Buffer has been optimized for use with TACS-Nuclease for the generation of a positive control for DNA fragmentation characteristic of apoptosis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 15 μL
Product Type TACS Nuclease and Buffer