missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Streptavidin-Alkaline Phosphatase

Product Code. 30021424
Change view
Click to view available options
:
1mL, Alkaline Phosphatase
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30021424 1mL, Alkaline Phosphatase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021424 Supplier R&D Systems™ Supplier No. AR001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: ELISA Detection (Matched Antibody Pair)

  • Results from the chemical conjugation of streptavidin and calf intestinal alkaline phosphatase
  • Recommended working dilution is 1:1000 in 1% BSA, 0.05% Tween™ 20, 0.025M Tris, 0.5M NaCl (pH 7.4)

This product contains sodium azide and thimerosal. Take appropriate protective precautions when working with these compounds.

Applications:

ELISA Detection

TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a solution in 0.1M Tris, 25mM MOPS, 2% polyethylene glycol, 50mM NaCl, 0.5mM Mg2Cl2, 0.005% thimerosal, 0.05% Tween 20, 0.75mM NaN3, 0.015 mM BSA.
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.