missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ StemXVivo Serum-Free Tumorsphere Media

Product Code. 18431150
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431150 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431150 Supplier R&D Systems™ Supplier No. CCM012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semisolid media formulated and optimized for tumorsphere formation

Tumorsphere assays, also called sphere-formation assays, are in vitro assays that assess stem cell characteristics of tumor cells. There are a number of variables that contribute to variation in data obtained from tumorsphere assays including the purity of the starting cell population, the type and quantity of media and supplements, and the potential for tumorspheres to aggregate.

  • Shown to support sphere formation of seven tumor cell lines
  • Formulated and optimized for efficient tumorsphere formation
  • Lot-to-lot consistency reduces experimental variability

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Media
Quantity 100 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.