missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RBC8

Product Code. 18741598
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18741598 10 mg 10mg
18794868 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18741598 Supplier Tocris Bioscience™ Supplier No. 5411/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

RalA and RalB GTPase inhibitor

RBC8 is a RalA and RalB GTPase inhibitor (EC50 ∽3.5 μM). Inhibits the binding of Ral to RaLBP1. Exhibits no detectable inhibition of Ras or RhoA activity. Suppresses growth of xenograft tumors in mice. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 361185-42-4
Synonym 6-Amino-4-(2,5-dimethoxyphenyl)-1,4-dihydro-3-(2-naphthalenyl)pyrano[2,3-c]pyrazole-5-carbonitrile
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C25H20N4O3
Formula Weight 424.45
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.