Learn More
R&D Systems™ Rat CD3+ T Cell Enrichment Columns
B cells bind to the anti-Ig coated beads and monocytes bind via Fc interactions for resulting T cell-rich eluate
Includes: Cell Selection Columns, 10X Column Buffer, Antibody Cocktail Vials
Description
Description
Purifies CD3+ T cells to 85 to 95%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Total CD3+ cell recovery ranges between 30% and 50% of initial CD3+ cells loaded;