Learn More
R&D Systems™ Proteome Profiler Human Pluripotent Stem Cell Array Kit
Product Code. 18037621
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18037621
Brand: R&D Systems™ ARY010
The Proteome Profiler Human Pluripotent Stem Cell Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Pluripotent Stem Cell Array Kit contains 8 membranes - each spotted in duplicate with 15 different stem cell marker antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit