Learn More
R&D Systems™ Proteome Profiler Human Apoptosis Array Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Proteome Profiler Human Apoptosis Array Kit contains 4 membranes - each spotted in duplicate with 35 different apoptosis antibodies.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 1 Kit |