Learn More
R&D Systems™ Proteome Profiler Human Angiogenesis Array Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
Description
The Proteome Profiler Human Angiogenesis Array Kit contains 4 membranes - each spotted in duplicate with 55 different angiogenesis antibodies.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 1 Kit |