missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PFK 15

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PFK 15 is an originally reported to be a selective PFKFB3 inhibitor (IC50 = 207 nM); a more recent report failed to demonstrate activity in a PFKFB3 kinase assay. Selective for PFKFB3 over a panel of 96 kinases. Reduces glucose uptake and induces apoptosis in H522 lung adenocarcinoma and Jurkat T cell leukemia cells. Suppresses glucose uptake and tumor growth, and inhibits metastasis of Lewis lung carcinoma cell xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | PFK 15 |
| CAS | 4382-63-2 |
| Synonym | 1-(4-Pyridinyl)-3-(2-quinolinyl)-2-propen-1-one |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C17H12N2O |
| Formula Weight | 260.29 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >99% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction