missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ONO 8130
GREENER_CHOICE

Product Code. 18778788
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18778788

Brand: Tocris Bioscience™ 5406/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EP1 receptor antagonist

ONO 8130 is an EP1 receptor antagonist. Exhibits analgesic effect in a mouse model of cystitis. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ONO 8130
CAS 459841-96-4
Synonym 4-[[[2,3-Dihydro-6-[(2-methylpropyl)[(4-methyl-2-thiazolyl)sulfonyl]amino]-1H-indene-5yl]oxy]methyl]benzoic acid
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C25H28N2O5S2
Formula Weight 500.63
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.