missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 8593 Hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.
Specifications
Specifications
| Chemical Name or Material | NS 8593 hydrochloride |
| CAS | 875755-24-1 |
| Synonym | N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Molecular Formula | C17H17N3·HCl |
| Formula Weight | 299.8 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction