missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 8593 Hydrochloride
GREENER_CHOICE

Product Code. 18750939
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18750939

Brand: Tocris Bioscience™ 4597/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective negative modulator of KCa2 (SK) channels; inhibits SK currents

NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NS 8593 hydrochloride
CAS 875755-24-1
Synonym N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C17H17N3·HCl
Formula Weight 299.8
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.