missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ N-2 Plus Media Supplement

Product Code. 18431940
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
18431940 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 18431940

Brand: R&D Systems™ AR003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

To culture neural progenitor cells and their differentiated derivatives under defined, serum-free conditions

Serum-free, defined media are routinely used as an alternative to standard serum-containing media in order to reduce unwanted experimental variability.

  • Chemically defined and serum-free
  • Consistent composition reduces experimental variation
  • Optimized for neural cell cultures
  • Contains high quality factors to support reproducible and efficient NPC expansion when combined with appropriate growth factors
  • N-2 Plus Media Supplement is a modification of Bottenstein's formulation
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type Media Supplement
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.