missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ N-2 MAX Media Supplement (100X)
Description
Serum-free, defined media are routinely used as an alternative to standard serum-containing media in order to reduce unwanted experimental variability.
- Contains high quality factors to support reproducible and efficient NPC expansion
- Is fully defined to reduce unwanted experimental variability
- Has been developed and optimized using neural progenitor cells
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 5 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?