missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse PTGFR (NP_032992) VersaClone cDNA
Beskrivning
The Mouse PTGFR (NP_032992) VersaClone cDNA clone contains the complete ORF for PTGFR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PTGFR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature.
Specifikationer
Specifikationer
| Gene | mPTGFR |
| Accession Number | NP_032992 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 1113bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?