missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse PlGF-2 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£285.09 - £812.07
Specifications
Gene ID (Entrez) | 18654 |
---|---|
Molecular Weight (g/mol) | 15.1 kDa (monomer) |
Name | PlGF-2 |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse PlGF-2 protein Ala24-Pro158 & Ala27-Pro158 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18157710
|
R&D Systems™
465-PL-010 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 10μg |
£285.09
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18303748
|
R&D Systems™
465-PL-010/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | 10μg |
£285.09
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18337437
|
R&D Systems™
465-PL-050 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 50μg |
£812.07
50µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18137350
|
R&D Systems™
465-PL-050/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | 50μg |
£812.07
50µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse VEGFR1/Flt-1 Fc Chimera (Catalog # 7756-FL ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Mouse PlGF 2 (Catalog # 465-PL) binds with an ED50 of 0.350-3.50 ng/mL.Specifications
18654 | |
PlGF-2 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
15.1 kDa (monomer) | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse PlGF-2 protein Ala24-Pro158 & Ala27-Pro158 | |
Disulfide-linked homodimer | |
Recombinant |