missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse MMP-7 Protein, CF

Product Code. 18329287
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329287 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329287 Supplier R&D Systems™ Supplier No. 2967MP010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (Catalog # ES010 ). The specific activity is >2000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA99983
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl, CaCl2 and Glycerol.
Gene ID (Entrez) 4316
Molecular Weight (g/mol) M.W. (Observed): 31 kDa, reducing conditions; M.W. (theoretical): 28 kDa
Name MMP-7
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse MMP-7 protein Met1-Leu264
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Pro form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Mmp7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein MMP-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.