missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse M-CSF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£269.97 - £787.24
Specifications
Gene ID (Entrez) | 12977 |
---|---|
Molecular Weight (g/mol) | 26 kDa (monomer) |
Name | M-CSF |
Source | E. coli-derived mouse M-CSF protein Lys33-Glu262,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18138590
|
R&D Systems™
416-ML-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 10μg |
£269.97
10µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18368397
|
R&D Systems™
416-ML-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 10μg |
£269.97
10µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18387427
|
R&D Systems™
416-ML-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 50μg |
£787.24
50µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18137760
|
R&D Systems™
416-ML-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 50μg |
£787.24
50µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Mouse M-CSF Protein is derived from E. coli. The Recombinant Mouse M-CSF Protein has been validated for the following applications: Bioactivity.Spécification
12977 | |
M-CSF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
26 kDa (monomer) | |
E. coli-derived mouse M-CSF protein Lys33-Glu262,with an N-terminal Met | |
Disulfide-linked homodimer | |
Recombinant |