missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse LPAR2/LPA2/EDG-4 (NP_064412) VersaClone cDNA

Product Code. 18441980
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18441980

Brand: R&D Systems™ RDC0626

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysophosphatidic Acid Receptor 2

The Mouse LPAR2/LPA2/EDG-4 (NP_064412) VersaClone cDNA clone contains the complete ORF for LPAR2/LPA2/EDG-4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LPAR2/LPA2/EDG-4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Gene mLPAR2
Accession Number NP_064412
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1060bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.