missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-5 ELISpot Kit

Product Code. 18752294
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18752294

Brand: R&D Systems™ EL405

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Cross Reactivity Mouse
Includes Mouse IL-5 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-5 Positive Control
Quantity 1 x 96-well Plate
Sample Type Whole cells
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Quantitative determination of the frequency of cells releasing mouse IL-5
Product Type Mouse IL-5 ELISpot Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.