Learn More
R&D Systems™ Mouse IL-5 ELISpot Development Module
Product Code. 18422250
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18422250 1 Kit 1 set
1 options
Product Code. 18422250 Supplier R&D Systems™ Supplier No. SEL405
Sufficient for at least five 96-well plates
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.