missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IFN-gamma ELISpot Kit

Product Code. 18772214
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18772214

Brand: R&D Systems™ EL485

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
TRUSTED_SUSTAINABILITY

Specifications

Includes One 96-well PVDF-backed plate coated w/mAb for mouse IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Quantity 1 x 96-well Plate
Sample Type Whole Cells
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Diagnostics
Product Type Mouse IFN-gamma ELISpot Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.