missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Granzyme B ELISpot Kit
Click to view available options
Quantity:
1 Kit
6 x 96-well Plates
Unit Size:
1 set
Specifications
Specifications
| Antigen | Granzyme B |
| Includes | One 96-well plate coated w/monoclonal Ab for mouse Granzyme B, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT chromogen, Positive control |
| Quantity | 1 Kit |
| Sample Type | Whole Cells |
| Synonym | C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Gene ID (Entrez) | 3002 |
| Gene Symbol | Gzmb |
| For Use With (Application) | Diagnostics |
| Host Species | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction