missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CTLA-4 Fc Chimera Protein

Product Code. 18366377
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
200µg
This item is not returnable. View return policy

Product Code. 18366377

Brand: R&D Systems™ 434CT200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit IL-2 secretion by stimulated Jurkat human acute T cell leukemia cells. Linsley, P.S. et al. (1991) J. Exp. Med. 174:561. The ED50 for this effect is 0.1-0.4 μg/mL when stimulated with 1 μg/mL Recombinant Human B7-1/CD80 Fc Chimera (Catalog # 140-B1 ) in the presence of PHA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6GTR6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1493
Molecular Weight (g/mol) M.W. (Observed): 55 kDa, reducing conditions; M.W. (theoretical): 41 kDa (monomer)
Name CTLA-4 Fc Chimera
Quantity 200 μg
Source Mouse myeloma cell line, NS0-derived mouse CTLA-4 protein Mouse CTLA-4 (Ala37-Phe162) (N-terminus) Accession #Q6GTR6 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Ctla4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CTLA-4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.