missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Crossveinless-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£369.32
Specifications
Gene ID (Entrez) | 73230 |
---|---|
Name | Crossveinless-2/CV-2 |
Quantity | 50μg |
Source | Mouse myeloma cell line,NS0-derived mouse Crossveinless-2/CV-2 protein Val34-Arg685,with an N-terminal 9-His tag,Ala39-Asp369 & Pro370-Arg685 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18118340
|
R&D Systems™
2299-CV-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£369.32
50µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18128180
|
R&D Systems™
2299-CV-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£369.32
50µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Crossveinless-2 Protein is derived from NS0. The Recombinant Mouse Crossveinless-2 Protein has been validated for the following applications: Bioactivity.Specifications
73230 | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Crossveinless-2/CV-2 | |
Mouse myeloma cell line,NS0-derived mouse Crossveinless-2/CV-2 protein Val34-Arg685,with an N-terminal 9-His tag,Ala39-Asp369 & Pro370-Arg685 | |
Disulfide-linked heterodimer | |
Recombinant |