missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Carboxypeptidase B1/CPB1 Protein, CF
Description
Measured by its ability to cleave a colorimetric peptide substrate, Hippuryl-Arg. The specific activity is >5,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_083982 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 1360 |
| Molecular Weight (g/mol) | M.W. (Observed): 43 kDa and 48 kDa, reducing conditions; M.W. (theoretical): 47 kDa |
| Name | Carboxypeptidase B1/CPB1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse Carboxypeptidase B1/CPB1 protein Ser16-Tyr415, with a C-terminal 10-His tag Accession # NP_083982 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?