missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Carboxypeptidase A4/CPA4 Protein, CF
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to cleave the colorimetric peptide substrate Ac-Phe-Thiaphe-OH in the presence of 5,5’Dithio-bis (2-nitrobenzoic acid) (DTNB). Edwards, K.M. et al. (1999) J. Biol. Chem. 274:30468. The specific activity is >400 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q6P8K8 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 51200 |
| Molecular Weight (g/mol) | M.W. (Observed): 48 kDa, reducing conditions; M.W. (theoretical): 47 kDa |
| Name | Carboxypeptidase A4/CPA4 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse Carboxypeptidase A4/CPA4 protein Gly17-Tyr420, with a C-terminal 10-His tag Accession # Q6P8K8 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction