missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMP-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£488.11
Specifications
Gene ID (Entrez) | 12162 |
---|---|
Molecular Weight (g/mol) | 15.6 kDa (monomer) |
Name | BMP-7 |
Quantity | 10μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse BMP-7 protein Ser292-His430 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18344827
|
R&D Systems™
5666-BP-010/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
£488.11
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18136800
|
R&D Systems™
5666-BP-010 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
£488.11
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse BMP-7 Protein is derived from CHO. The Recombinant Mouse BMP-7 Protein has been validated for the following applications: Bioactivity.Specifications
12162 | |
BMP-7 | |
Chinese Hamster Ovary cell line,CHO-derived mouse BMP-7 protein Ser292-His430 | |
Disulfide-linked homodimer | |
Recombinant |
15.6 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |