missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse ACE-2 Protein, CF

Product Code. 18337737
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337737 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337737 Supplier R&D Systems™ Supplier No. 3437ZN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >400 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_081562
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and ZnCl2.
Gene ID (Entrez) 59272
Molecular Weight (g/mol) M.W. (Observed): 110 kDa, reducing conditions; M.W. (theoretical): 85 kDa
Name ACE-2
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse ACE-2 protein Gln18-Thr740, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Recombinant Mouse ACE-2 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Ace2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ACE-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.