missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
Description
Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.008-0.05 μg/mL in the presence of a Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ). Measured by its ability to bind Recombinant Mouse 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 937-4B ) in a functional ELISA.
Specifications
Specifications
| Accession Number | P41274 |
| For Use With (Application) | Bioactivity, Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 8744 |
| Molecular Weight (g/mol) | M.W. (Observed): 41-44 kDa, reducing conditions; M.W. (theoretical): 25.7 kDa |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse 4-1 BB Ligand/TNFSF9 protein Met 10-His tag GGGSGGGSGGGSIEGR Mouse 4-1 BB Ligand (Arg104-Glu309) (N-terminus) Accession # P41274 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Symbol | Tnfsf9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction