missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein
Description
Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.008-0.05 μg/mL in the presence of a Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ). Measured by its ability to bind Recombinant Mouse 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 937-4B ) in a functional ELISA.
Specifications
Specifications
| Accession Number | P41274 |
| For Use With (Application) | Bioactivity, Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 8744 |
| Molecular Weight (g/mol) | M.W. (Observed): 41-44 kDa, reducing conditions; M.W. (theoretical): 25.7 kDa |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse 4-1 BB Ligand/TNFSF9 protein Met 10-His tag GGGSGGGSGGGSIEGR Mouse 4-1 BB Ligand (Arg104-Glu309) (N-terminus) Accession # P41274 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Symbol | Tnfsf9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?