missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 179
GREENER_CHOICE

Product Code. 18794529
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18794529

Brand: Tocris Bioscience™ 4957/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective liver receptor homolog 1 (LRH1) inverse agonist

ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ML 179
CAS 1883548-87-5
Synonym 3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C21H25F3N4O2
Formula Weight 422.44
Physical Form Crystalline Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.