missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MK6-83
GREENER_CHOICE

Product Code. p-7082199
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18703439

Brand: Tocris Bioscience™ 5547/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRPML channel activator

MK6-83 is a TRPML channel activator (EC50 = 110 nM). Also activates F408Δ and F465L TRPML mutants (EC50 values are 0.1 and 1.23 μM respectively). Restores endolysosomal trafficking and zinc homeostasis in lysosomes of mucolipidosis type IV (MLIV) mutant fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MK6-83
CAS 1062271-24-2
Synonym 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C16H20N2O2S2
Formula Weight 336.47
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.