missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MK6-83
GREENER_CHOICE

Product Code. 18756529
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18756529 50 mg 50mg
18703439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18756529 Supplier Tocris Bioscience™ Supplier No. 5547/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRPML channel activator

MK6-83 is a TRPML channel activator (EC50 = 110 nM). Also activates F408Δ and F465L TRPML mutants (EC50 values are 0.1 and 1.23 μM respectively). Restores endolysosomal trafficking and zinc homeostasis in lysosomes of mucolipidosis type IV (MLIV) mutant fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MK6-83
CAS 1062271-24-2
Synonym 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C16H20N2O2S2
Formula Weight 336.47
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.