missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (-)-JQ1
Description
(-)-JQ1 is an inactive control for (+)-JQ1 . Exhibits no significant interaction with BRD1-4 or a panel of other bromodomains.
Active Enantiomer also available.
Specifications
Specifications
| Chemical Name or Material | (-)-JQ1 |
| CAS | 1268524-71-5 |
| Synonym | (6R)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetic acid 1,1-dimethylethyl ester |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C23H25ClN4O2S |
| Formula Weight | 456.99 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?