missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronan (Low Molecular Weight), R&D Systems™
Specifications
Specifications
| Chemical Name or Material | Hyaluronan |
| Color | White |
| Physical Form | White odorless powder |
| Synonym | Hyaluronic acid (HA), Sodium hyaluronate |
| Quantity | 100 mg |
| Formula Weight | Predicted MW: 15 - 40kDa Observed MW: 20.0kDa |
| Odor | Odorless |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?