missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TMEM106B (NP_001127704) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Beskrivning
The Human TMEM106B (NP_001127704) VersaClone cDNA clone contains the complete ORF for TMEM106B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TMEM106B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature.
Specifikationer
Specifikationer
| Gene | hTMEM106B |
| Accession Number | NP_001127704 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 837bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering