missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human SGLT1/SLC5A1 (NP_000334) VersaClone cDNA

Product Code. 18491290
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18491290 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18491290 Supplier R&D Systems™ Supplier No. RDC0654
  • ONLINE EXCLUSIVE PRICE
    £693.00
    £655.20 / 10µg
    Save £37.80
    5% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Sodium/Glucose Transporter 1

The Human SGLT1/SLC5A1 (NP_000334) VersaClone cDNA clone contains the complete ORF for SGLT1/SLC5A1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SGLT1/SLC5A1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifikationer

Gene hSLC5A1
Accession Number NP_000334
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 2008bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.