missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Proinsulin Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
Includes
Human proinsulin microplate, 96-well; Human proinsulin conjugate; Human proinsluin standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers
The Human Proinsulin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Proinsulin in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | HRP |
| Assay Sensitivity | 1.43 pM |
| Assay Range | 3.28 to 210 pM |
| For Use With (Application) | ELISA |
| Sample Type | Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL) |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction