missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human OX40/TNFRSF4 Fc Chimera Protein, CF

Product Code. 18393728
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393728 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393728 Supplier R&D Systems™ Supplier No. 3388OX050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human OX40/TNFRSF4 Fc Chimera is immobilized at 10 ng/mL, the concentration of Recombinant Human OX40 Ligand/TNFSF4 (Catalog # 1054-OX ) that produces 50% optimal binding response is found to be approximately 0.5-3 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43489
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7293
Molecular Weight (g/mol) M.W. (Observed): 70 kDa, reducing conditions; M.W. (theoretical): 46.7 kDa (monomer)
Name OX40/TNFRSF4 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human OX40/TNFRSF4 protein Human OX40 (Leu29-Ala216) (N-terminus) Accession # P43489 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFRSF4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein OX40/TNFRSF4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.